Sample vials
24,00 ml, 23×86 mm, Glas, klar, No, N-20
Katalog # WITG9.350.306
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Sample vials made of borosilicate glass for general use.
Specifikationer
- Kasket inkluderet:No
- Cap størrelse:N-20
- Diameter×H:23×86 mm
- Kapacitet:24,00 ml
- Farve:klar
- Materiale:Glas