Sample vials
8,00 ml, 17×61 mm, Glas, klar, No, N-15
Catalog # WITG9.350.303
Supplier: witeg Labortechnik
Gå til:
Product Details & Documents
Sample vials made of borosilicate glass for general use.
Specifikationer
- Kasket inkluderet:No
- Cap størrelse:N-15
- Diameter×H:17×61 mm
- Kapacitet:8,00 ml
- Farve:klar
- Materiale:Glas