Natriumhydroxid ≥99.99% (metals basis), beads/pellets
Glass bottle for solids, 100 g
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
45780.22
: 1310-73-2
Purity: 99.99% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Density: 2.13 Sensitivity: Air Sensitive and Hygroscopic Appearance: White to off-white Odour: Odourless pH: 14 Melting Point: 316° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :14,8627
- :1390 °C (1013 hPa)
- :316 °C
- :2,13
- :Stue temperatur
- :215-185-5
- :1823
- :8,II
- :01-2119457892-27
Documents
- Pakning:Glass bottle for solids
- Pakn:100 g