Orders LinkContactUsLinkComponent
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
Glass bottle for solids, 100 g
  45780.22
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
  45780.22
 :  1310-73-2
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
Glass bottle for solids, 100 g
 :  1310-73-2
1.760,00 kr
1 * 100 g
Purity: 99.99% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Density: 2.13 Sensitivity: Air Sensitive and Hygroscopic Appearance: White to off-white Odour: Odourless pH: 14 Melting Point: 316° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548
 :  Caustic soda
  •  :1310-73-2
  •  :NaOH
  •  :MFCD00003548
  •  :40 g/mol
  •  :14,8627
  •  :1390 °C (1013 hPa)
  •  :316 °C
  •  :2,13
  •  :Stue temperatur
  •  :215-185-5
  •  :1823
  •  :8,II
  •  :01-2119457892-27
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g