Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumhydroxid ≥99.99% (metals basis), beads/pellets

Glass bottle for solids, 5 g
Katalog # 45780.06
Ætsende
Leverandør:   Thermo Fisher Scientific
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
Katalog # 45780.06
CAS nummer:  1310-73-2
Natriumhydroxid ≥99.99% (metals basis), beads/pellets
Natriumhydroxid ≥99.99% (metals basis), beads/pellets

Glass bottle for solids, 5 g

CAS nummer:  1310-73-2
Price
418,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Purity: 99.99% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Density: 2.13 Sensitivity: Air Sensitive and Hygroscopic Appearance: White to off-white Odour: Odourless pH: 14 Melting Point: 316° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548

Synonymer:  Caustic soda
  • CAS nummer:1310-73-2
  • Formel:NaOH
  • MDL nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,8627
  • Kogende Pt:1390 °C (1013 hPa)
  • Smeltende Pt:316 °C
  • Tæthed:2,13
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-185-5
  • FN:1823
  • ADR:8,II
  • NÅ:01-2119457892-27
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer