Stands for MSC-Advantage
Manually adjustable floor stand, 750 to 950 mm working height in 50 mm increments, for 0,9 m cabinet
Katalog # 135-1320
Leverandør: Thermo Scientific
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Manually adjustable floor stand, 750 to 950 mm working height in 50 mm increments, for 0,9 m cabinet
- Seq:05