Orders LinkContactUsLinkComponent
Reducing screw thread adaptor couplings, BOLA
Reducing screw thread adaptor couplings, BOLA
GL 25 and GL 18
Katalog # BOHLH904-04
Leverandør:   Bohlender
Reducing screw thread adaptor couplings, BOLA
Katalog # BOHLH904-04
Leverandør:  Bohlender
Reducing screw thread adaptor couplings, BOLA
Reducing screw thread adaptor couplings, BOLA
GL 25 and GL 18
Price
491,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Connector, reducing screw thread coupling, made of PTFE. Two screw caps made of PPS with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
  • Applicable for connecting two GL threads of different sizes
  • Temperature resisitance from −200 to +250 ºC
  • Very good chemical resistance
  • FDA conformity
Specifikationer
  • Gevind:
    GL 25 and GL 18