Reducing screw thread adaptor couplings, BOLA
GL 25 and GL 18
Katalog # BOHLH904-04
Leverandør: Bohlender
Gå til:
Product Details & Documents
Connector, reducing screw thread coupling, made of PTFE. Two screw caps made of PPS with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
- Applicable for connecting two GL threads of different sizes
- Temperature resisitance from −200 to +250 ºC
- Very good chemical resistance
- FDA conformity
Specifikationer
- Gevind:GL 25 and GL 18