Geraniol 99%
Glass bottle, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: (E)-3,7-Dimethylocta-2,6-dien-1-ol
- CAS nummer:106-24-1
- Formel:C₁₀H₁₈O
- MDL nummer:MFCD00002917
- Molecular Weight:154.25 g/mol
- Kogende Pt:229…230 °C (757 mmHg)
- Smeltende Pt:–15 °C
- Tæthed:0,878 g/cm³
- Flash Pt:76 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-377-1
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4780 (20°C, 589 nm)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



