Dropping bottles, glass
100 ml, DIN 18
Katalog # HECH41314060
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Made of amber glass.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Amber coloured to protect light sensitive substances
Dropping bottles without cap, with thread DIN 18.
Bestillingsoplysninger: Pipette kit and screw cap must be ordered seperately.
Specifikationer
- Gevind:DIN 18
- Kapacitet:100 ml
- Seq:05
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 5 ml dropping bottlesPipette kit, red, for 5 ml dropping bottlesHECH41315006Log ind for tilgængelighed1 * 100 Items - 662,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, blue, for 20 ml dropping bottlesPipette kit, blue, for 20 ml dropping bottlesHECH41315020Log ind for tilgængelighed1 * 100 Items - 643,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, blue, for 30 ml dropping bottlesPipette kit, blue, for 30 ml dropping bottlesHECH41315030Log ind for tilgængelighed1 * 100 Items - 662,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 30 ml dropping bottlesPipette kit, red, for 30 ml dropping bottlesHECH41315031Log ind for tilgængelighed1 * 100 Items - 662,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 100 ml dropping bottlesPipette kit, red, for 100 ml dropping bottlesHECH41315061Log ind for tilgængelighed1 * 100 Items - 674,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 20 ml dropping bottlesPipette kit, red, for 20 ml dropping bottlesHECH41315021Log ind for tilgængelighed1 * 100 Items - 643,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 10 ml dropping bottlesPipette kit, red, for 10 ml dropping bottlesHECH41315011Log ind for tilgængelighed1 * 100 Items - 643,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 50 ml dropping bottlesPipette kit, red, for 50 ml dropping bottlesHECH41315051Log ind for tilgængelighed1 * 100 Items - 653,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, blue, for 50 ml dropping bottlesPipette kit, blue, for 50 ml dropping bottlesHECH41315650Log ind for tilgængelighed1 * 100 Items - 466,00 kr