Orders LinkContactUsLinkComponent
Dropping funnels, spherical, without pressure balance
Dropping funnels, spherical, without pressure balance
500 ml, 29/32 NS, 29/32 NS
Catalog # WITG1.892.500
Supplier:   witeg Labortechnik
Dropping funnels, spherical, without pressure balance
Catalog # WITG1.892.500
Supplier:  witeg Labortechnik
Dropping funnels, spherical, without pressure balance
Dropping funnels, spherical, without pressure balance
500 ml, 29/32 NS, 29/32 NS
Price
1.180,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Dropping funnels, spherical, made of borosilicate glass 3.3, stopcock with glass key. With NS cone and socket.
  • Without graduations
  • Excellent chemical resistance
  • Excellent resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    500 ml