Dropping funnels, spherical, without pressure balance
500 ml, 29/32 NS, 29/32 NS
Catalog # WITG1.892.500
Supplier: witeg Labortechnik
Dropping funnels, spherical, without pressure balance
Catalog # WITG1.892.500
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Dropping funnels, spherical, made of borosilicate glass 3.3, stopcock with glass key. With NS cone and socket.
- Without graduations
- Excellent chemical resistance
- Excellent resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
- Konusstr.:29/32 NS
- Kappestr.:29/32 NS
- Kapacitet:500 ml