Dropping funnels, spherical, without pressure balance
100 ml, 29/32 NS, 29/32 NS
WITG1.892.100
Dropping funnels, spherical, made of borosilicate glass 3.3, stopcock with glass key. With NS cone and socket.
- Without graduations
- Excellent chemical resistance
- Excellent resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
- Konusstr.:29/32 NS
- Kappestr.:29/32 NS
- Kapacitet:100 ml