Orders LinkContactUsLinkComponent
Dropping funnels, spherical, without pressure balance
100 ml, 29/32 NS, 29/32 NS
  WITG1.892.100
Dropping funnels, spherical, without pressure balance
  WITG1.892.100
Dropping funnels, spherical, without pressure balance
Dropping funnels, spherical, without pressure balance
100 ml, 29/32 NS, 29/32 NS
747,00 kr
1 * 1 Items
Dropping funnels, spherical, made of borosilicate glass 3.3, stopcock with glass key. With NS cone and socket.
  • Without graduations
  • Excellent chemical resistance
  • Excellent resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    100 ml