Xylitol 99+%
Plastic bottle, 25 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
- CAS nummer:87-99-0
- Formel:C₅H₁₂O₅
- MDL nummer:MFCD00064292
- Molecular Weight:152.15 g/mol
- Merck Index:13,10140
- Kogende Pt:215…217 °C
- Smeltende Pt:92…96 °C
- Tæthed:1,52 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-788-0
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:92 to 96 °C
- Water:≤0.5 %
- Sulfated ash:≤0.5 %
- GC:≤2.0 % Other polyols
- Arsenic (As):≤3 ppm
- Heavy metals (as Pb):≤10 ppm
- Reducing sugars:≤0.2 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:25 g
- Cat. No.:ACRO225980250



