Orders LinkContactUsLinkComponent
Penetration probes, K-Type
Penetration probes, K-Type
Spring-loaded V4A tube with Ø 1,5 mm slim insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
Response time (T90): water 0,4 m/s ~3 s, −65…+550 °C
Katalog # GRSG600514
Leverandør:   Greisinger
Penetration probes, K-Type
Katalog # GRSG600514
Leverandør:  Greisinger
Penetration probes, K-Type
Penetration probes, K-Type
Spring-loaded V4A tube with Ø 1,5 mm slim insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
Response time (T90): water 0,4 m/s ~3 s, −65…+550 °C
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Område:
    −65…+550 °C
  • Beskrivelse:
    Spring-loaded V4A tube with Ø 1,5 mm slim insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
    Response time (T90): water 0,4 m/s ~3 s
  • Seq:
    02