Cover glasses, square
10×10 mm, #1, Borosilikatglas
Katalog # HECH40990101
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Borosilicate glass coverslips for histology and microscopy.
Specifikationer
- Antal:#1
- L×B:10×10 mm
- Materiale:Borosilikatglas