Active gauge displays
Active gauge displays for APG-L with control relays (mbar/Torr)
EDWAD39561000
The active gauge displays are a range of simple, low cost units for the measurement and control of vacuum from atmosphere to 10-9 mbar.
- Single switch with LED indication for gauge head selection
- Option of two adjustable set-points with relay status indication
- Mains power input supply via IEC connector
- Clear analog display covers the whole gauge head pressure range
- Available with both bench top use or panel mounting
: Supplied with tilt feet for convenient bench top mounting and panel mounting kit to DIN 43700 standard.
- Beskrivelse:Active gauge displays for APG-L with control relays (mbar/Torr)
- Seq:01
- Cat. No.:EDWAD39561000
- Effekt, W:90 to 120 V ac
185 to 265 V ac (selectable) - Tilladt driftstemperatur:5...+60 °C
- Model:APG-L
- Tilladt rumtemperatur under opbevaring:−10...+70 °C