Orders LinkContactUsLinkComponent
Sample bottles, glass
Sample bottles, glass
250 ml, Prøveudtagningsflaske, glas, 250 ml, GL 45
Catalog # 300-0055
Supplier:   BURKLE
Sample bottles, glass
Catalog # 300-0055
Supplier:  BURKLE
Sample bottles, glass
Sample bottles, glass
250 ml, Prøveudtagningsflaske, glas, 250 ml, GL 45
Price
302,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Specifications
  • Gevind:
    GL 45
  • Kapacitet:
    250 ml
  • Beskrivelse:
    Prøveudtagningsflaske, glas, 250 ml
  • Seq:
    02