Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Total with Neutr/ Staph, 120
Catalog # LIOF53530
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF53530
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides
Total with Neutr/ Staph, 120
CAS Number:  
Price
2.210,00 kr
Log in for availability
1 * 120 Items
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Specifications
Specifications
  • Pakn:
    120
  • Beskrivelse:
    Total with Neutr/ Staph