Pipette tips
Eppendorf type, 5 - 200 µl, +, Yellow, Eppendorf, Gilson, Socorex, Nichiryo, Biohit, Brand, Individually wrapped
Leverandør: DELTALAB
Gå til:
Product Details & Documents
Documents
Specifikationer
- Pakket:Individually wrapped
- Steril:+
- Volumen:5 - 200 µl
- Seq:10
- Spidstype:Eppendorf type
- Spits farge:Yellow
- Til brug med:Eppendorf, Gilson, Socorex, Nichiryo, Biohit, Brand