Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HyperSep™ Hypercarb™ SPE columns

HyperSep™ Hypercarb™ SPE columns

1 ml, 100 mg
Katalog # 549-0622
Leverandør:   Thermo Fisher Scientific
HyperSep™ Hypercarb™ SPE columns
Katalog # 549-0622
HyperSep™ Hypercarb™ SPE columns
HyperSep™ Hypercarb™ SPE columns

1 ml, 100 mg

Price
5.160,00 kr
Log ind for tilgængelighed
1 * 30 Items
Product Details & Documents

Flat, 100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents. Hydrophobic reversed phase; normal phase adsorption; polar retention.

  • Provides total pH stability and the retention and separation of highly polar species where silica and resin columns fail
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • No restriction on eluents
Specifikationer
  • Kapacitet:
    1 ml
  • Seq:
    03
  • Lejevægt:
    100 mg