Orders LinkContactUsLinkComponent
HyperSep™ Hypercarb™ SPE columns
6 ml, 1000 mg
  549-0626
HyperSep™ Hypercarb™ SPE columns
  549-0626
HyperSep™ Hypercarb™ SPE columns
HyperSep™ Hypercarb™ SPE columns
6 ml, 1000 mg
8.940,00 kr
1 * 10 Items
Flat, 100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents. Hydrophobic reversed phase; normal phase adsorption; polar retention.
  • Provides total pH stability and the retention and separation of highly polar species where silica and resin columns fail
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • No restriction on eluents
  • Kapacitet:
    6 ml
  • Seq:
    06
  • Lejevægt:
    1000 mg