Orders LinkContactUsLinkComponent
Stands for MSC-Advantage
Stands for MSC-Advantage
Fixed height floor stand, 750 mm working height, for 1,5 m cabinet
Katalog # 135-1319
Leverandør:   Thermo Scientific
Stands for MSC-Advantage
Katalog # 135-1319
Leverandør:  Thermo Scientific
Stands for MSC-Advantage
Stands for MSC-Advantage
Fixed height floor stand, 750 mm working height, for 1,5 m cabinet
Price
8.670,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Fixed height floor stand, 750 mm working height, for 1,5 m cabinet
  • Seq:
    04