PTFE balls, BOLA
Balls, PTFE, 15 mm
BOHLH964-15
Made from solid PTFE, with smooth surface.
For extension of surfaces of e.g. distillation apparatus; as splash guard.
- Ø:15 mm
- Beskrivelse:Balls
- Materiale:PTFE
- Seq:008