[EN]SEMI-MICRO CELL 10MM LID QUARTZ 1 * 2 Items
Katalog # PERCB0631049
Leverandør: PerkinElmer
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Leverandør: VWR Chemicals
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
VWR®, Ansigtsservietter
Leverandør: VWR Collection
Hydrogenperoxid 30%, (Perhydrol®) EMSURE® ISO for analysis, Supelco®
Leverandør: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
VWR®, Nitril handsker, NITRILE
Leverandør: VWR Collection
VWR®, Short Thread Vials ND9
Leverandør: VWR Collection
Vand, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverandør: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Leverandør: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Leverandør: VWR Collection
pH-elektroder, InLab® ISM
Leverandør: Mettler - Toledo
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals