Orders LinkContactUsLinkComponent
Microsampling tools
Microsampling tools
Pin vise
Katalog # PERCN9302603
Leverandør:   PerkinElmer
Microsampling tools
Katalog # PERCN9302603
Leverandør:  PerkinElmer
Microsampling tools
Pin vise
Price
395,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This microsampling tool kit is suitable for FTIR microscopy.
Leveringsinformation: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.
Specifikationer
  • Beskrivelse:
    Pin vise
  • Seq:
    06