Orders LinkContactUsLinkComponent
Interference check standards for ICP-OES
Interference check standards for ICP-OES
Interferenskontrolstandard 5, 5% HNO₃, 6.000 mg/l: Ca; 5.000 mg/l: Fe; 3.000 mg/l: Mg; 1.200 mg/l: Al; 1.000 mg/l: Na, 1
Katalog # PERCN9300208
Leverandør:   PerkinElmer
Interference check standards for ICP-OES
Katalog # PERCN9300208
Leverandør:  PerkinElmer
CAS nummer:  
Interference check standards for ICP-OES
Interferenskontrolstandard 5, 5% HNO₃, 6.000 mg/l: Ca; 5.000 mg/l: Fe; 3.000 mg/l: Mg; 1.200 mg/l: Al; 1.000 mg/l: Na, 1
CAS nummer:  
Price
3.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
Specifikationer
  • Matrice:
    5% HNO₃
  • Pakn:
    1
  • Beskrivelse:
    Interferenskontrolstandard 5
  • Indhold:
    6.000 mg/l: Ca; 5.000 mg/l: Fe; 3.000 mg/l: Mg; 1.200 mg/l: Al; 1.000 mg/l: Na