Blades
Barberblade, enkelt kant, ryg i aluminium, 100 blade pr. æske, pakket enkeltvis, Kulstofstål, 0,22 mm, 38,1×19 mm
Catalog # PERS94-0462
Supplier: PERSONNA MEDICAL
Jump to:
Product Details & Documents
Single-use blades for a variety of applications.
Specifications
- Beskrivelse:Barberblade, enkelt kant, ryg i aluminium, 100 blade pr. æske, pakket enkeltvis
- L×B:38,1×19 mm
- Materiale:Kulstofstål
- Seq:06
- Tykkelse:0,22 mm