Digestion tubes, DigiTUBE®
62,5 ml, Borosilikatglas, Tube, calibrated, without caps and rack lock
PERCN9308050
Disposable, sample digestion tubes with class A graduations.
Rack lock design allows for easy, one handed closure of leak free screw cap.
- Kapacitet:62,5 ml
- Beskrivelse:Tube, calibrated, without caps and rack lock
- Materiale:Borosilikatglas
- Seq:10
Accessories
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse