Orders LinkContactUsLinkComponent
Digestion tubes, DigiTUBE®
Digestion tubes, DigiTUBE®
62,5 ml, Borosilikatglas, Tube, calibrated, without caps and rack lock
  PERCN9308050
Digestion tubes, DigiTUBE®
  PERCN9308050
Digestion tubes, DigiTUBE®
62,5 ml, Borosilikatglas, Tube, calibrated, without caps and rack lock
4.070,00 kr
1 * 1 Items
Disposable, sample digestion tubes with class A graduations.
Rack lock design allows for easy, one handed closure of leak free screw cap.
  • Kapacitet:
    62,5 ml
  • Beskrivelse:
    Tube, calibrated, without caps and rack lock
  • Materiale:
    Borosilikatglas
  • Seq:
    10
Accessories
  •  
    Beskrivelse
  • PERCN9308705
    Screw cap, blue, for DigiTube®
    1 * 540 Items
  •  
    Beskrivelse
  • PERCN9308708
    Screw cap, clear, for DigiTube®
    1 * 540 Items - 785,00 kr
  •  
    Beskrivelse
  • PERCN9308027
    Screw cap, PTFE, for PTFE DigiTube®
    1 * 6 Items - 7.740,00 kr
  •  
    Beskrivelse
  • PERCN9308058
    Screw cap, orange, for DigiTube®
    1 * 250 Items - 235,00 kr
  •  
    Beskrivelse
  • PERCN9308059
    Screw cap, red, for DigiTube®
    1 * 250 Items - 370,00 kr
  •  
    Beskrivelse
  • PERCN9308060
    Screw cap, clear, for DigiTube®
    1 * 250 Items - 1.480,00 kr