Digestion tubes, DigiTUBE®
62,5 ml, Kvarts, Tube, uncalibrated, without caps and rack lock
Catalog # PERCN9308048
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Disposable, sample digestion tubes with class A graduations.
Rack lock design allows for easy, one handed closure of leak free screw cap.
Specifications
- Kapacitet:62,5 ml
- Beskrivelse:Tube, uncalibrated, without caps and rack lock
- Materiale:Kvarts
- Seq:08
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice