Interference check standards for ICP-OES
Analyt C-opløsning, 2 mg/L: Ag, As, Ba, Be, Cd, Co, Cr, Cu, Hg*, Mn, Ni, Pb, Sb, Se, Tl, V, Zn, 1
PERCN9303831
- Pakn:1
- Beskrivelse:Analyt C-opløsning
- Indhold:2 mg/L: Ag, As, Ba, Be, Cd, Co, Cr, Cu, Hg*, Mn, Ni, Pb, Sb, Se, Tl, V, Zn