Orders LinkContactUsLinkComponent
Interference check standards for ICP-OES
Interferenskontrolopløsning 2, 2% HNO₃, 10 mg/l: Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn, 1
Catalog # PERCN9303830
Supplier:   PerkinElmer
Interference check standards for ICP-OES
Catalog # PERCN9303830
Supplier:  PerkinElmer
CAS Number:  
Interference check standards for ICP-OES
Interferenskontrolopløsning 2, 2% HNO₃, 10 mg/l: Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn, 1
CAS Number:  
Price
2.420,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
Specifications
  • Matrice:
    2% HNO₃
  • Pakn:
    1
  • Beskrivelse:
    Interferenskontrolopløsning 2
  • Indhold:
    10 mg/l: Ag, As, Cd, Co, Cr, Cu, Mn, Ni, Zn