Orders LinkContactUsLinkComponent
Interference check standards for ICP-OES
Interference check standards for ICP-OES
Interferenskontrolopløsning 1, 10.000 mg/l: Cl; 2.000 mg/l: C; 1.000 mg/l: Al, Ca, Fe, K, Mg, Na, P, S; 20 mg/l: Mo, Ti, 1
  PERCN9303828
Interference check standards for ICP-OES
  PERCN9303828
 :  
Interference check standards for ICP-OES
Interferenskontrolopløsning 1, 10.000 mg/l: Cl; 2.000 mg/l: C; 1.000 mg/l: Al, Ca, Fe, K, Mg, Na, P, S; 20 mg/l: Mo, Ti, 1
 :  
6.290,00 kr
1 * 1 Items
  • Pakn:
    1
  • Beskrivelse:
    Interferenskontrolopløsning 1
  • Indhold:
    10.000 mg/l: Cl; 2.000 mg/l: C; 1.000 mg/l: Al, Ca, Fe, K, Mg, Na, P, S; 20 mg/l: Mo, Ti