Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

7-Bromisatin ≥97%

Glass bottle for solids, 1 g
Katalog # H32843.03
Irriterende
Leverandør:   Thermo Fisher Scientific
7-Bromisatin ≥97%
Katalog # H32843.03
CAS nummer:  20780-74-9
7-Bromisatin ≥97%
7-Bromisatin ≥97%

Glass bottle for solids, 1 g

CAS nummer:  20780-74-9
Price
397,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 20780-74-9 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Melting Point: 195-199° MDL No.: MFCD00774354

Synonymer:  7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione
  • CAS nummer:20780-74-9
  • Formel:C₈H₄BrNO₂
  • MDL nummer:MFCD00774354
  • Molecular Weight:226.03 g/mol
  • Smeltende Pt:195…199 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer