Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
7-Bromisatin ≥97%

7-Bromisatin ≥97%

Glass bottle for solids, 10 g
Katalog # H32843.09
Irriterende
Leverandør:   Thermo Fisher Scientific
7-Bromisatin ≥97%
Katalog # H32843.09
CAS nummer:  20780-74-9
7-Bromisatin ≥97%
7-Bromisatin ≥97%

Glass bottle for solids, 10 g

CAS nummer:  20780-74-9
Price
2.240,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents

Purity: 97% CAS No.: 20780-74-9 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Melting Point: 195-199° MDL No.: MFCD00774354

Synonymer:  7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione
  • CAS nummer:20780-74-9
  • Formel:C₈H₄BrNO₂
  • MDL nummer:MFCD00774354
  • Molecular Weight:226.03 g/mol
  • Smeltende Pt:195…199 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.