Orders LinkContactUsLinkComponent
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Single channel pipettes, mechanical, fixed / variable volume, labopette®
5 µl, ±2,5 %, <1,5 %, Clear, 0,2 - 10 µl
Catalog # HIRS9475501
Supplier:   HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Catalog # HIRS9475501
Supplier:  HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Single channel pipettes, mechanical, fixed / variable volume, labopette®
5 µl, ±2,5 %, <1,5 %, Clear, 0,2 - 10 µl
Price
1.970,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The reliable labopette® pipette features superior ergonomic characteristics, right down to the smallest detail. The entire working process is optimised to ensure non fatiguing work that places a minimum of stress on the joints.
  • Volume configuration with digital locking feature
  • With tip ejection and calibration option
  • Tip seal and safety filter to prevent contamination
  • Trigger grip ensures a firm but relaxed hold during pipetting
  • Ergonomic design of the handle offers an equally firm, comfortable hold for large and small hands, right and left-handed users and hands with or without gloves
  • Effective thermal insulation of the unit interior prevents hand heat penetrating into the interior of the unit
  • Completely autoclavable without dismantling
 : Supplied with calibration tool, pipette holder, individual serial number and calibration certificate.
Specifications
  • Volumen:
    5 µl
  • Seq:
    09
  • Nøjagtighed:
    ±2,5 %
  • Usikkerhed:
    <1,5 %
  • Til pipettespidser:
    Clear, 0,2 - 10 µl
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9470010
    Safe-Cone-filtre, til labopette® variabel 1.000 - 5.000 µl; fast 2.000/5.000 µl
    Log in for availability
    1 * 50 Items - 462,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9470013
    Safe-Cone-filtre, til labopette® variabel 2 - 20/5 - 50 µl; fast 20/25/50 µl
    Log in for availability
    1 * 50 Items - 462,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9470011
    Safe-Cone-filtre, til labopette® elektronisk 10 - 500/50 - 1.000/500 - 5.000/50 - 1200 µl; variabel 100 - 1.000 µl; fast 250/500/1.000 µl
    Log in for availability
    1 * 50 Items - 462,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9470012
    Safe-Cone-filtre, til labopette® variabel 10 - 100/20 - 200 µl; fast 100/200 µl
    Log in for availability
    1 * 50 Items - 462,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9980116
    DKD-kalibreringscertifikat til labopette® fast 0,5 μl - 5 ml
    Log in for availability
    1 * 1 Items - 680,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9980118
    DKD-kalibreringscertifikat til labopette® 12-kanal, 5 - 250 μl
    Log in for availability
    1 * 1 Items - 2.950,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9470006
    Stativ til seks mekaniske pipetter
    Log in for availability
    1 * 1 Items - 679,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9470004
    Karruselstativ til seks mekaniske pipetter
    Log in for availability
    1 * 1 Items - 1.380,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490101
    0,2 - 10 µl
    Transparent
    Hirschmann Laborgeräte, Gilson®-pipetter
    Log in for availability
    1 * 1.000 Items - 267,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490102
    1 - 200 µl
    Yellow
    Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter
    Log in for availability
    1 * 1.000 Items - 178,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490103
    100 - 1000 µl
    Blue
    Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter
    Log in for availability
    1 * 1.000 Items - 198,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490106
    1 - 250 µl
    Transparent
    labopette®-multikanalpipetter
    Log in for availability
    1 * 1.000 Items - 289,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490124
    100 - 5000 µl
    Transparent
    Hirschmann Laborgeräte-pipetter
    Log in for availability
    1 * 300 Items - 423,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490201
    0,2 - 10 µl
    Transparent
    Hirschmann Laborgeräte, Gilson®-pipetter
    Log in for availability
    1 * 960 Items - 635,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490202
    1 - 200 µl
    Yellow
    Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter
    Log in for availability
    1 * 960 Items - 479,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490205
    50 - 1200 µl
    Transparent
    Hirschmann Laborgeräte-pipetter
    Log in for availability
    1 * 960 Items - 1.190,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490206
    1 - 250 µl
    Transparent
    labopette®-multikanalpipetter
    Log in for availability
    1 * 960 Items - 476,00 kr
Product Information
  • Catalog #
    Volumen
    Spits farge
    Til brug med
    Availability
    Price
  • HIRS9490104
    1 - 10000 µl
    Transparent
    Hirschmann Laborgeräte-pipetter
    Log in for availability
    1 * 100 Items - 423,00 kr
Product Information
  • Catalog #
    Spidstype
    Til brug med
    Availability
    Price
  • HIRS9490109
    Multibox, autoclavable
    refill trays of pipette tips up to 300 µl
    Log in for availability
    1 * 1 Items - 227,00 kr
Product Information
  • Catalog #
    Spidstype
    Til brug med
    Availability
    Price
  • HIRS9490110
    Multibox, autoclavable
    refill trays of pipette tips up to 1000 µl
    Log in for availability
    1 * 1 Items - 240,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9470014
    Safe-Cone-filtre, til labopette® fast 10/20/25/250 µl; flerkanals 5,0 - 50/50 - 300 µl; elektronisk flerkanals 25 - 250/5 - 100 µl
    Log in for availability
    1 * 50 Items - 462,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9980115
    DKD-kalibreringscertifikat til labopette® variable volumener 0,5 μl - 5 ml/labopettor
    Log in for availability
    1 * 1 Items - 1.100,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HIRS9980117
    DKD-kalibreringscertifikat til labopette® 8-kanal, 0,5 - 250 μl
    Log in for availability
    1 * 1 Items - 2.630,00 kr