Drenchers
Drencher, straight, 102 mm, 1,6 mm
Catalog # SOCO376.16102S
Supplier: Socorex Isba
Gå til:
Product Details & Documents
These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.
- Luer lock fittings
Specifikationer
- Udv.Ø:1,6 mm
- Beskrivelse:Drencher, straight
- Længde:102 mm
- Seq:07