Pipette tips
VistaTip™, pyrgen-free, 1250 µl, +, Individually wrapped
Catalog # VIST2028
Supplier: VISTALAB TECHNOLOGIES
Jump to:
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifications
- Pakket:Individually wrapped
- Steril:+
- Volumen:1250 µl
- Seq:10
- Spidstype:VistaTip™, pyrgen-free