Orders LinkContactUsLinkComponent
Single channel pipettes, electronic, variable volume, Ovation®
Model for left handers, 25 - 1250 µl, 5 µl, ±2,8 - ±0,75 %, <0,60 - <0,15 %, lilla
Katalog # VIST1065-1250L
Leverandør:   VISTALAB TECHNOLOGIES
Single channel pipettes, electronic, variable volume, Ovation®
Katalog # VIST1065-1250L
Leverandør:  VISTALAB TECHNOLOGIES
Single channel pipettes, electronic, variable volume, Ovation®
Single channel pipettes, electronic, variable volume, Ovation®
Model for left handers, 25 - 1250 µl, 5 µl, ±2,8 - ±0,75 %, <0,60 - <0,15 %, lilla
Price
6.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These pipettes are the ideal ergonomic choice for users who need a comprehensive range of automated liquid handling functions. An easy-to-read display and simple-to-operate keypad allows fast recall and setup of user-defined pipetting protocols and selection of operating modes, volumes and speeds.
  • Fully electronic operation maximises pipetting efficiency for tasks like multiple dispensing, pipetting, reverse pipetting, mixing, serial diluting and automatic sequential dispensing
  • Colour-coded right and left-handed models are available
 : These models require a power supply unit (sold separately) to recharge the pipette's battery.
Specifikationer
  • Farvekode:
    lilla
  • Volumen:
    25 - 1250 µl
  • Beskrivelse:
    Model for left handers
  • Seq:
    08
  • Forøgelse:
    5 µl
  • Nøjagtighed:
    ±2,8 - ±0,75 %
  • Usikkerhed:
    <0,60 - <0,15 %
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Power supply, 90-264 VAC, 47-63 Hz, for one Ovation® pipette
    Power supply, 90-264 VAC, 47-63 Hz, for one Ovation® pipette
    VIST9060-9005
    Log ind for tilgængelighed
    1 * 1 Items - 404,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Power supply, with four connections, for up to four Ovation® pipettes
    Power supply, with four connections, for up to four Ovation® pipettes
    VIST9060-9006
    Log ind for tilgængelighed
    1 * 1 Items - 1.250,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement nozzle for ESC model (0,5 - 20 µl); QS and M models (1 - 10/2 - 20 µl); F1 and F2 models (3 - 20 µl)
    Replacement nozzle for ESC model (0,5 - 20 µl); QS and M models (1 - 10/2 - 20 µl); F1 and F2 models (3 - 20 µl)
    613-4946
    Log ind for tilgængelighed
    1 * 10 Items - 660,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 0,2 - 2 µl (QS), Ovation 1 and 2 µl (F1 and F2)
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 0,2 - 2 µl (QS), Ovation 1 and 2 µl (F1 and F2)
    VIST9057-1001
    Log ind for tilgængelighed
    1 * 1 Each - 218,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 1 - 10 µl (QS), Ovation 3 - 10 µl (F1 and F2)
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 1 - 10 µl (QS), Ovation 3 - 10 µl (F1 and F2)
    VIST9057-1002
    Log ind for tilgængelighed
    1 * 1 Each - 218,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    VIST9057-2035
    Log ind for tilgængelighed
    1 * 1 Each - 282,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    VIST9057-3036
    Log ind for tilgængelighed
    1 * 1 Each - 282,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 10 - 100 µl (QS), Ovation 21 - 100 µl (F1 and F2)
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 10 - 100 µl (QS), Ovation 21 - 100 µl (F1 and F2)
    VIST9057-2004
    Log ind for tilgængelighed
    1 * 1 Each - 218,00 kr