Orders LinkContactUsLinkComponent
Single channel pipettes, electronic, variable volume, Ovation®
Model for right handers, 2 - 125 µl, 0,5 µl, ±2,9 - ±0,8 %, <1,00 - <0,15 %, grøn
  VIST1065-0125
Single channel pipettes, electronic, variable volume, Ovation®
  VIST1065-0125
Single channel pipettes, electronic, variable volume, Ovation®
Single channel pipettes, electronic, variable volume, Ovation®
Model for right handers, 2 - 125 µl, 0,5 µl, ±2,9 - ±0,8 %, <1,00 - <0,15 %, grøn
6.280,00 kr
1 * 1 Items
These pipettes are the ideal ergonomic choice for users who need a comprehensive range of automated liquid handling functions. An easy-to-read display and simple-to-operate keypad allows fast recall and setup of user-defined pipetting protocols and selection of operating modes, volumes and speeds.
  • Fully electronic operation maximises pipetting efficiency for tasks like multiple dispensing, pipetting, reverse pipetting, mixing, serial diluting and automatic sequential dispensing
  • Colour-coded right and left-handed models are available
 : These models require a power supply unit (sold separately) to recharge the pipette's battery.
  • Farvekode:
    grøn
  • Volumen:
    2 - 125 µl
  • Beskrivelse:
    Model for right handers
  • Seq:
    04
  • Forøgelse:
    0,5 µl
  • Nøjagtighed:
    ±2,9 - ±0,8 %
  • Usikkerhed:
    <1,00 - <0,15 %
Accessories
  •  
    Beskrivelse
  • VIST9060-9005
    Power supply, 90-264 VAC, 47-63 Hz, for one Ovation® pipette
    1 * 1 Items - 404,00 kr
  •  
    Beskrivelse
  • VIST9060-9006
    Power supply, with four connections, for up to four Ovation® pipettes
    1 * 1 Items - 1.250,00 kr
  •  
    Beskrivelse
  • 613-4946
    Replacement nozzle for ESC model (0,5 - 20 µl); QS and M models (1 - 10/2 - 20 µl); F1 and F2 models (3 - 20 µl)
    1 * 10 Items - 660,00 kr
  •  
    Beskrivelse
  • VIST9057-1001
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 0,2 - 2 µl (QS), Ovation 1 and 2 µl (F1 and F2)
    1 * 1 Each - 218,00 kr
  •  
    Beskrivelse
  • VIST9057-1002
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 1 - 10 µl (QS), Ovation 3 - 10 µl (F1 and F2)
    1 * 1 Each - 218,00 kr
  •  
    Beskrivelse
  • VIST9057-2035
    VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    1 * 1 Each - 282,00 kr
  •  
    Beskrivelse
  • VIST9057-3036
    VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    1 * 1 Each - 282,00 kr
  •  
    Beskrivelse
  • VIST9057-2004
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 10 - 100 µl (QS), Ovation 21 - 100 µl (F1 and F2)
    1 * 1 Each - 218,00 kr