Orders LinkContactUsLinkComponent
Spatulas
Spatulas
240 mm, 30 mm, hvid
Catalog # COWI043.240
Supplier:   COWIE
Spatulas
Catalog # COWI043.240
Supplier:  COWIE
Spatulas
Spatulas
240 mm, 30 mm, hvid
Price
235,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Double-ended spatula/scraper combination made of PTFE, with fine chamfered ends.
Documents
Specifications
  • Farve:
    hvid
  • Længde:
    240 mm
  • Bladbredde:
    30 mm