Spatulas
210 mm, 25 mm, hvid
Catalog # COWI043.210
Supplier: COWIE
Jump to:
Product Details & Documents
Double-ended spatula/scraper combination made of PTFE, with fine chamfered ends.
Documents
COWI043.240_TDS_EN
Datasheet
Specifications
- Farve:hvid
- Længde:210 mm
- Bladbredde:25 mm