L(-)-Carnitine
1 Vial
Katalog # CAMBNSK-B-1
Leverandør: CAMBRIDGE ISOTOPE LABS
Gå til:
Product Details & Documents
Synonymer: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS nummer:541-15-1
- Formel:C₇H₁₅NO₃
- MDL nummer:MFCD00038747
- Molecular Weight:161.20 g/mol
- EINECS:208-768-0
Specifikationer
Specifikationer
- Pakn:1 Vial
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



