Orders LinkContactUsLinkComponent
Stands for MSC-Advantage
Stands for MSC-Advantage
Fixed height floor stand, 750 mm working height, for 0,9 m cabinet
Katalog # 135-1318
Leverandør:   Thermo Scientific
Stands for MSC-Advantage
Katalog # 135-1318
Leverandør:  Thermo Scientific
Stands for MSC-Advantage
Stands for MSC-Advantage
Fixed height floor stand, 750 mm working height, for 0,9 m cabinet
Price
7.240,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Fixed height floor stand, 750 mm working height, for 0,9 m cabinet
  • Seq:
    01