Orders LinkContactUsLinkComponent
Skuffe racks til cryobokse
15 (5×3), 140×421×285 mm
Catalog # 479-0760
Supplier:   TENAK
Skuffe racks til cryobokse
Catalog # 479-0760
Supplier:  TENAK
Skuffe racks til cryobokse
Skuffe racks til cryobokse
15 (5×3), 140×421×285 mm
Price
1.470,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Rustfrit stål.
  • Racks til 5 forskellige højder æsker
  • Kan bruges i alle typer frysere
  • Passer til alle standard cryobokse
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility. Measure the inside height and depth of your freezer in order to choose the right rack.
 : Delivered with or without cryoboxes.
Specifications
  • Rum:
    15 (5×3)
  • B×D×H udv.:
    140×421×285 mm
  • Seq:
    19