Screw caps
S 28
Catalog # VITL102697
Supplier: VITLAB
Jump to:
Product Details & Documents
PFA, transparent
- Especially suitable for use in trace analysis
- Ideal for sensitive and valuable samples
- High thermal stability, from –200 °C to +260 °C, autoclavable
- Chemical inertness against nearly all chemicals
- Easy to clean
For sealing all PFA containers with GL threads or buttress threads.
Specifications
- Gevind:S 28