Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Viscosity standards, general purpose

Viscosity standards, general purpose

S30000, Glass bottle, 1
Katalog # CANN9727-C65
Viscosity standards, general purpose
Katalog # CANN9727-C65
Leverandør:  
CAS nummer:  
Viscosity standards, general purpose

S30000, Glass bottle, 1

CAS nummer:  
Discontinued
Product Details & Documents

These certified standards are useful for calibration and verification procedures.

  • It is suitable for all types of viscometers
  • Viscosity data quoted at 20 °C
  • It also helps ensures the reliability of measurements

Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1
  • Antal:
    S30000

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.