Industrial Standing Rests
Industrial ESD standing rest 2, with a swivel range of 360⁰, 650 - 850 mm, Flat disc base
Catalog # 132-1421
Supplier: bimos
Jump to:
Product Details & Documents
Industrial standing rest supports the body and relieve the strain from the spinal column.
- Seat made from integral foam and with integrated carry handle
- Seat can be tilted forwards by 10°
Specifications
- Beskrivelse:Industrial ESD standing rest 2, with a swivel range of 360⁰
- Højde:650 - 850 mm
- Seq:02
- Størrelse:Flat disc base