Industrial Standing Rests
Industrial standing rest 2, with pneumatic spring, swivel range of 360⁰, 650 - 850 mm, Disc base
Katalog # 132-1420
Leverandør: bimos
Gå til:
Product Details & Documents
Industrial standing rest supports the body and relieve the strain from the spinal column.
- Seat made from integral foam and with integrated carry handle
- Seat can be tilted forwards by 10°
Specifikationer
- Beskrivelse:Industrial standing rest 2, with pneumatic spring, swivel range of 360⁰
- Højde:650 - 850 mm
- Seq:01
- Størrelse:Disc base