Brushes for test tubes
Test tube brush, with fallow bristles, Hog bristle, 105 mm, 26 mm, Wire, 230 mm
Katalog # REIT12240301
Leverandør: REITENSPIESS BUERSTEN
Gå til:
Product Details & Documents
Brushes with natural bristles and zinc plated wire handle.
Specifikationer
- Børstemateriale:Hog bristle
- Børstehoved Ø:26 mm
- Handle material:Wire
- Beskrivelse:Test tube brush, with fallow bristles
- Seq:20
- Samlet længde:230 mm
- Børstehovedets længde:105 mm