Corning® Cap, Phenolic, PTFE Liner
24-410, Reusable phenolic screw cap, PTFE liner, sort
Supplier: Corning
Corning® Cap, Phenolic, PTFE Liner
Catalog # CORN9998-241
Supplier: Corning
Jump to:
Product Details & Documents
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
- Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
Documents
Specifications
- Gevind:24-410
- med:PTFE liner
- Farve:sort
- Beskrivelse:Reusable phenolic screw cap
- Seq:05
- Cat. No.:CORN9998-241
- Miljøcertificering:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Lav fremstillingspåvirkning