Orders LinkContactUsLinkComponent
Corning® Cap, Phenolic, PTFE Liner
Corning® Cap, Phenolic, PTFE Liner
24-410, Reusable phenolic screw cap, PTFE liner, sort
Catalog # CORN9998-241
   Lav fremstillingspåvirkning
Supplier:   Corning
Corning® Cap, Phenolic, PTFE Liner
Catalog # CORN9998-241
Supplier:  Corning
   Lav fremstillingspåvirkning
Corning® Cap, Phenolic, PTFE Liner
Corning® Cap, Phenolic, PTFE Liner
24-410, Reusable phenolic screw cap, PTFE liner, sort
Price
2.470,00 kr
Log in for availability
1 * 144 Items
Product Details & Documents
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
  • Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
Specifications
  • Gevind:
    24-410
  • med:
    PTFE liner
  • Farve:
    sort
  • Beskrivelse:
    Reusable phenolic screw cap
  • Seq:
    05
  • Cat. No.:
    CORN9998-241
  • Miljøcertificering:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Lav fremstillingspåvirkning