Orders LinkContactUsLinkComponent
Corning® Cap, Phenolic, PTFE Liner
Corning® Cap, Phenolic, PTFE Liner
18-415, Reusable phenolic screw cap, PTFE-pakning, sort
Katalog # 212-7529
   Lav fremstillingspåvirkning
Leverandør:   Corning
Corning® Cap, Phenolic, PTFE Liner
Katalog # 212-7529
Leverandør:  Corning
   Lav fremstillingspåvirkning
Corning® Cap, Phenolic, PTFE Liner
Corning® Cap, Phenolic, PTFE Liner
18-415, Reusable phenolic screw cap, PTFE-pakning, sort
Price
1.040,00 kr
Log ind for tilgængelighed
1 * 192 Items
Product Details & Documents
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
  • Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
Specifikationer
  • Gevind:
    18-415
  • med:
    PTFE-pakning
  • Farve:
    sort
  • Beskrivelse:
    Reusable phenolic screw cap
  • Seq:
    03
  • Cat. No.:
    212-7529
  • Miljøcertificering:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Lav fremstillingspåvirkning