Corning® Cap, Phenolic, PTFE Liner
18-415, Reusable phenolic screw cap, PTFE-pakning, sort
Leverandør: Corning
Corning® Cap, Phenolic, PTFE Liner
Katalog # 212-7529
Leverandør: Corning
Gå til:
Product Details & Documents
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
- Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
Documents
Specifikationer
- Gevind:18-415
- med:PTFE-pakning
- Farve:sort
- Beskrivelse:Reusable phenolic screw cap
- Seq:03
- Cat. No.:212-7529
- Miljøcertificering:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Lav fremstillingspåvirkning