Accessories for multi-parameter meters, MultiLine® IDS
QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)
Leverandør: WTW
Gå til:
Product Details & Documents
Documents
Certifikater
Søg efter partinummerinoLab® Multi IDS.pdf Sensors for Field and Laboratory.pdf
Additional Product Details
Specifikationer
- Beskrivelse:QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)
- Seq:34