Orders LinkContactUsLinkComponent
Accessories for multi-parameter meters, MultiLine® IDS
Accessories for multi-parameter meters, MultiLine® IDS
QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)
Katalog # 662-2132
Leverandør:   WTW
Accessories for multi-parameter meters, MultiLine® IDS
Katalog # 662-2132
Leverandør:  WTW
Accessories for multi-parameter meters, MultiLine® IDS
Accessories for multi-parameter meters, MultiLine® IDS
QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)
Price
392,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    QSC kit, initial calibration kit for IDS pH electrodes (Box with 3 ampoules: pH 4,01/pH 6,86/pH 9,18)
  • Seq:
    34